Assay Details
Target Gene Details
Entrez Gene ID: | 12563 |
Gene Name: | cadherin 6 |
Gene Aliases: |
K-cadherin, cad6, cadherin-6 |
Location: |
Chr.15:13028699-13173689 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh6 | NM_007666.3 | NP_031692.2 | ||
BC107008.1 | AAI07009.1 | |||
BC107013.1 | AAI07014.1 | |||
D82029.1 | BAA11516.1 | |||
U67399.1 | AAB07550.1 |
More Information
Set Membership: |
Intragenic Non-exonic |