Assay Details
Target Gene Details
Entrez Gene ID: | 12465 |
Gene Name: | chaperonin containing Tcp1, subunit 5 (epsilon) |
Gene Aliases: |
Ccte, TCPE, mKIAA0098 |
Location: |
Chr.15:31590813-31601804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct5 | NM_007637.2 | NP_031663.1 | ||
AK077753.1 | ||||
AK088184.1 | BAC40194.1 | |||
AK129056.1 | BAC97866.1 | |||
AK133857.1 | BAE21891.1 | |||
AK136493.1 | BAE23007.1 | |||
AK145445.1 | BAE26441.1 | |||
AK146405.1 | BAE27146.1 | |||
AK146425.1 | BAE27159.1 | |||
AK150390.1 | BAE29518.1 | |||
AK153911.1 | BAE32250.1 | |||
AK167895.1 | BAE39906.1 | |||
BC094427.1 | AAH94427.1 | |||
Z31555.1 | CAA83430.1 |
More Information
Set Membership: |
Intragenic Non-exonic |