Assay Details
Target Gene Details
Entrez Gene ID: | 268780 |
Gene Name: | EGF-like, fibronectin type III and laminin G domains |
Gene Aliases: |
5930412K08, AU040377 |
Location: |
Chr.15:7206120-7398746 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egflam | NM_001289496.1 | 6 | 975 | NP_001276425.1 |
NM_001289498.1 | 6 | 970 | NP_001276427.1 | |
NM_178748.6 | 6 | 975 | NP_848863.1 | |
AK031136.1 | 6 | 884 | BAC27271.1 | |
AK033332.1 | 6 | 892 | BAC28235.1 | |
AK037223.1 | 6 | 877 | BAC29762.1 | |
AK041546.1 | 6 | 856 | BAC30982.1 | |
AK087659.1 | 6 | 855 | ||
AK158909.1 | 6 | 971 | ||
BC095994.1 | 6 | 705 | AAH95994.1 | |
BC150710.1 | 6 | 695 | ||
DQ223720.1 | 6 | 679 |
More Information
Set Membership: |
Intragenic Exonic |