Assay Details
Target Gene Details
Entrez Gene ID: | 67040 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 |
Gene Aliases: |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
Location: |
Chr.15:79527696-79546764 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx17 | NM_001040187.1 | 2 | 587 | NP_001035277.1 |
NM_199079.1 | 2 | 406 | NP_951061.1 | |
NM_199080.2 | 2 | 587 | NP_951062.1 | |
AK039888.1 | 2 | 406 | BAC30474.1 | |
AK151061.1 | 2 | 362 | BAE30078.1 | |
AK152749.1 | 2 | 362 | ||
AK152831.1 | 2 | 362 | BAE31531.1 | |
AK161001.1 | 2 | 403 | ||
AK161012.1 | 2 | 402 | BAE36146.1 | |
AK191519.1 | ||||
BC062910.1 | 2 | 392 | AAH62910.1 | |
BC096036.1 | 2 | 587 | AAH96036.1 |