Assay Details
Target Gene Details
Entrez Gene ID: | 18163 |
Gene Name: | catenin (cadherin associated protein), delta 2 |
Gene Aliases: |
Catnd2, Nprap, neurojugin |
Location: |
Chr.15:30172593-31029344 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnnd2 | NM_008729.2 | 21 | 3870 | NP_032755.2 |
AK140454.1 | 11 | 1488 | BAE24395.1 | |
AK140960.1 | 6 | 745 | BAE24530.1 | |
AK164175.1 | 2 | 343 | ||
AK164304.1 | 4 | 421 | BAE37727.1 | |
BC138260.1 | 21 | 3415 | ||
BC138261.1 | 21 | 3415 | ||
BC145217.1 | 20 | 3337 | ||
U90331.1 | 21 | 3873 | AAB82409.1 |
More Information
Set Membership: |
Intragenic Exonic |