Assay Details
Target Gene Details
Entrez Gene ID: | 14042 |
Gene Name: | exostoses (multiple) 1 |
Gene Aliases: |
AA409028 |
Location: |
Chr.15:53068261-53346183 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ext1 | NM_010162.2 | 6 | 2296 | NP_034292.2 |
AK132327.1 | 6 | 2273 | BAE21106.1 | |
BC004741.1 | 6 | 1660 | AAH04741.1 | |
U78539.1 | 6 | 1800 | AAB41728.1 | |
X96639.1 | 6 | 2070 | CAA65443.1 |
More Information
Set Membership: |
Intragenic Exonic |