Assay Details
Target Gene Details
Entrez Gene ID: | 67040 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 |
Gene Aliases: |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
Location: |
Chr.15:79527696-79546764 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx17 | NM_001040187.1 | NP_001035277.1 | ||
NM_152806.3 | 1 | 1044 | NP_690019.2 | |
NM_199080.2 | NP_951062.1 | |||
AK151061.1 | BAE30078.1 | |||
AK152749.1 | ||||
AK152831.1 | BAE31531.1 | |||
AK152910.1 | BAE31589.1 | |||
AK161012.1 | 12 | 2859 | BAE36146.1 | |
BC027758.1 | 1 | 281 | AAH27758.2 | |
BC038378.1 | 1 | 630 | AAH38378.1 | |
BC096036.1 | AAH96036.1 | |||
BI655302.1 |