Assay Details
Target Gene Details
Entrez Gene ID: | 13196 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain1 |
Gene Aliases: |
AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19 |
Location: |
Chr.15:64086840-64382919 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asap1 | NM_001276461.1 | 5 | 562 | NP_001263390.1 |
NM_001276462.1 | 3 | 471 | NP_001263391.1 | |
NM_001276463.1 | 3 | 471 | NP_001263392.1 | |
NM_001276467.1 | 5 | 562 | NP_001263396.1 | |
NM_010026.3 | 5 | 562 | NP_034156.2 | |
AF075461.1 | 5 | 381 | AAC98349.1 | |
AF075462.1 | 5 | 381 | AAC98350.1 | |
AK122477.1 | 3 | 166 | BAC65759.1 | |
AK132979.1 | 3 | 301 | ||
AK146545.1 | 5 | 400 | BAE27250.1 | |
AK147689.1 | 3 | 472 | ||
BC048818.1 | 3 | 437 | AAH48818.1 | |
BC094581.1 | 5 | 562 | AAH94581.1 | |
CB527210.1 | 3 | 265 | ||
CD355387.1 | 1 | 59 | ||
CF735731.1 |