Assay Details
Target Gene Details
Entrez Gene ID: | 66168 |
Gene Name: | glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding) |
Gene Aliases: |
1110025J15Rik, Lag, Tmbim3 |
Location: |
Chr.15:76246740-76249911 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grina | NM_023168.3 | NP_075657.1 | ||
AF182040.1 | AAG16897.1 | |||
AK003920.1 | ||||
AK179160.1 | ||||
AK189399.1 | ||||
AK200011.1 | ||||
AK212726.1 | ||||
BC010802.1 | AAH10802.1 | |||
BC019157.1 | AAH19157.1 | |||
BC037667.1 | AAH37667.2 |
Target Gene Details
Entrez Gene ID: | 102465575 |
Gene Name: | microRNA 6953 |
Gene Aliases: |
Gm27738, mmu-mir-6953 |
Location: |
Chr.15:76248191-76248258 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir6953 | NR_105918.1 |