Assay Details
Target Gene Details
Entrez Gene ID: | 100503915 |
Gene Name: | sphingomyelin phosphodiesterase 5 |
Gene Aliases: |
ENSMUSG00000071724, Gm10345, MA-nSMase |
Location: |
Chr.15:76293869-76296896 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpd5 | NM_001195537.1 | NP_001182466.1 | ||
AK005822.1 | ||||
BY705974.1 | ||||
CF197814.1 | ||||
GU144514.1 |