Assay Details
Target Gene Details
Entrez Gene ID: | 328572 |
Gene Name: | E1A binding protein p300 |
Gene Aliases: |
A430090G16, A730011L11, KAT3B, p300, p300 HAT |
Location: |
Chr.15:81586214-81652077 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ep300 | NM_177821.6 | 3 | 1191 | NP_808489.4 |
AK042627.1 | 3 | 1187 | BAC31311.1 | |
BC144976.1 | 3 | 1045 | ||
BC150681.1 | 3 | 1045 |
More Information
Set Membership: |
Intragenic Exonic |