Assay Details
Target Gene Details
Entrez Gene ID: | 77627 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
4931407K02Rik, 4932408N08Rik, bM150J22.2, mKIAA1672 |
Location: |
Chr.15:83866712-84075025 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efcab6 | NM_001161628.1 | NP_001155100.1 | ||
NM_001161629.1 | NP_001155101.1 | |||
NM_029946.4 | NP_084222.4 | |||
AK016516.1 | ||||
AK019850.1 | BAB31880.1 | |||
AK220412.1 | BAD90458.1 | |||
BC056955.1 | AAH56955.1 | |||
BC065111.1 | AAH65111.1 |
More Information
Set Membership: |
Intragenic Non-exonic |