Assay Details
Target Gene Details
Entrez Gene ID: | 77627 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
4931407K02Rik, 4932408N08Rik, bM150J22.2, mKIAA1672 |
Location: |
Chr.15:83866712-84075025 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efcab6 | NM_001161628.1 | 3 | 435 | NP_001155100.1 |
NM_029946.4 | 20 | 2542 | NP_084222.4 | |
AK019850.1 | 3 | 435 | BAB31880.1 | |
AK220412.1 | 3 | 354 | BAD90458.1 | |
BC056955.1 | 8 | 945 | AAH56955.1 | |
BC065111.1 | 20 | 2542 | AAH65111.1 |
More Information
Set Membership: |
Intragenic Exonic |