Assay Details
Target Gene Details
Entrez Gene ID: | 12983 |
Gene Name: | colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage) |
Gene Aliases: |
AI848964, AIC2B, Bc, CDw131, Csf2rb1, Csfgmrb, Il3r, Il3rb, Il3rb1, Il5rb |
Location: |
Chr.15:78325797-78351001 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csf2rb | NM_007780.4 | NP_031806.3 | ||
AF483496.1 | AAL90770.1 | |||
AF483497.1 | AAL90771.1 | |||
AK152608.1 | BAE31354.1 | |||
AK154286.1 | ||||
AK155109.1 | BAE33054.1 | |||
AK155215.1 | BAE33126.1 | |||
AK155699.1 | BAE33394.1 | |||
AK155803.1 | BAE33441.1 | |||
AK155822.1 | ||||
AK155885.1 | ||||
AK171477.1 | BAE42480.1 | |||
BC113202.1 | AAI13203.1 | |||
M34397.1 | AAA37204.1 | |||
S78451.1 | AAB34209.1 |
More Information
Set Membership: |
Intragenic Non-exonic |