Assay Details
Target Gene Details
Entrez Gene ID: | 21816 |
Gene Name: | transglutaminase 1, K polypeptide |
Gene Aliases: |
2310004J08Rik, Tgase1 |
Location: |
Chr.14:55700009-55713921 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgm1 | NM_001161714.1 | NP_001155186.1 | ||
NM_001161715.1 | NP_001155187.1 | |||
NM_019984.3 | NP_064368.3 | |||
AF186373.1 | AAF35986.1 | |||
BC026422.1 | AAH26422.1 |
More Information
Set Membership: |
Intragenic Non-exonic |