Assay Details
Target Gene Details
Entrez Gene ID: | 75288 |
Gene Name: | solute carrier family 35, member F4 |
Gene Aliases: |
4930550L21Rik |
Location: |
Chr.14:49298519-49525837 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc35f4 | NM_029238.2 | NP_083514.1 | ||
AB041538.1 | BAA95023.1 | |||
AK034308.1 | BAC28670.1 | |||
BC138981.1 | ||||
BC145720.1 |