Assay Details
Target Gene Details
Entrez Gene ID: | 382913 |
Gene Name: | nei like 2 (E. coli) |
Gene Aliases: |
Gm1212, NEH2 |
Location: |
Chr.14:63182445-63194244 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Neil2 | NM_201610.2 | 2 | 188 | NP_963904.2 |
AK155596.1 | 2 | 337 | ||
AY518221.1 | 2 | 204 | AAR98807.1 | |
BC115709.1 | 2 | 188 | ||
BC115762.1 | 2 | 188 | AAI15763.1 |
More Information
Set Membership: |
Intragenic Exonic |