Assay Details
Target Gene Details
Entrez Gene ID: | 18300 |
Gene Name: | oncoprotein induced transcript 1 |
Gene Aliases: |
2310076N21Rik, AV067083, EF-7, Fam3d |
Location: |
Chr.14:8348937-8378763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oit1 | NM_146050.2 | NP_666162.1 | ||
AK009397.1 | ||||
AK010206.1 | BAC25286.1 | |||
BC024504.1 | AAH24504.1 | |||
BC024532.1 | AAH24532.1 | |||
BC024542.1 | AAH24542.1 | |||
BC024636.1 | AAH24636.1 | |||
BC034193.1 | AAH34193.1 | |||
U72677.2 | AAB51037.1 |
More Information
Set Membership: |
Intragenic Non-exonic |