Assay Details
Target Gene Details
Entrez Gene ID: | 68646 |
Gene Name: | NAD kinase 2, mitochondrial |
Gene Aliases: |
MNADK, Nadkd1 |
Location: |
Chr.15:9071260-9110496 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nadk2 | NM_001040395.4 | NP_001035485.2 | ||
NM_001085410.2 | NP_001078879.1 | |||
NM_001286253.1 | NP_001273182.1 | |||
NM_001286255.1 | NP_001273184.1 | |||
AK003858.1 | BAB23041.1 | |||
AK016988.1 | BAB30539.1 | |||
AK046666.1 | ||||
AK078318.1 | BAC37218.1 | |||
AK149267.1 | BAE28780.1 | |||
AK162601.1 | BAE36982.1 | |||
BC115776.1 |