Assay Details
Target Gene Details
Entrez Gene ID: | 74551 |
Gene Name: | phosphoenolpyruvate carboxykinase 2 (mitochondrial) |
Gene Aliases: |
1810010O14Rik, 9130022B02Rik, PEPCK-M |
Location: |
Chr.14:55540263-55550777 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pck2 | NM_028994.2 | NP_083270.1 | ||
AK007429.1 | ||||
AK030501.1 | BAC26991.1 | |||
AK031612.1 | BAC27477.1 | |||
AK034927.1 | BAC28883.1 | |||
AK147972.1 | BAE28259.1 | |||
AK165094.1 | ||||
AK170242.1 | BAE41656.1 | |||
BC010318.1 | AAH10318.1 | |||
BC023437.1 | AAH23437.1 |
More Information
Set Membership: |
Intragenic Non-exonic |