Assay Details
Target Gene Details
Entrez Gene ID: | 74551 |
Gene Name: | phosphoenolpyruvate carboxykinase 2 (mitochondrial) |
Gene Aliases: |
1810010O14Rik, 9130022B02Rik, PEPCK-M |
Location: |
Chr.14:55540263-55550777 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pck2 | NM_028994.2 | 3 | 579 | NP_083270.1 |
AK030501.1 | 3 | 580 | BAC26991.1 | |
AK031612.1 | 3 | 424 | BAC27477.1 | |
AK034927.1 | 3 | 458 | BAC28883.1 | |
AK147972.1 | 3 | 433 | BAE28259.1 | |
AK165094.1 | 3 | 457 | ||
AK170242.1 | 3 | 433 | BAE41656.1 | |
AK200018.1 | ||||
BC010318.1 | 3 | 442 | AAH10318.1 | |
BC023437.1 | 1 | 47 | AAH23437.1 |
More Information
Set Membership: |
Intragenic Exonic |