Assay Details
Target Gene Details
Entrez Gene ID: | 70646 |
Gene Name: | N(alpha)-acetyltransferase 30, NatC catalytic subunit |
Gene Aliases: |
4930487N19Rik, 5730533P17Rik, AI447922, AW322455, Nat12 |
Location: |
Chr.14:49172133-49191031 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naa30 | NM_001081430.1 | NP_001074899.1 | ||
AK015640.1 | BAC25468.1 | |||
AK039333.1 | ||||
AK044235.1 | ||||
BC054060.1 | AAH54060.1 | |||
BC157931.1 | ||||
BC158065.1 | ||||
BC172029.1 |
More Information
Set Membership: |
Intragenic Non-exonic |