Assay Details
Target Gene Details
Entrez Gene ID: | 22334 |
Gene Name: | voltage-dependent anion channel 2 |
Gene Aliases: |
Vdac6, mVDAC2, mVDAC6 |
Location: |
Chr.14:21831561-21845879 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vdac2 | NM_011695.2 | NP_035825.1 | ||
AK012359.1 | BAE43231.1 | |||
AK078555.1 | ||||
AK150504.1 | BAE29617.1 | |||
AK150627.1 | BAE29717.1 | |||
AK150940.1 | BAE29975.1 | |||
AK152274.1 | BAE31090.1 | |||
AK159561.1 | BAE35185.1 | |||
AK168199.1 | BAE40159.1 | |||
AK168328.1 | ||||
BC003731.1 | AAH03731.2 | |||
U30838.1 | AAC13321.1 |
More Information
Set Membership: |
Intragenic Non-exonic |