Assay Details
Target Gene Details
Entrez Gene ID: | 219024 |
Gene Name: | transmembrane protein 55b |
Gene Aliases: |
BC039161 |
Location: |
Chr.14:50926070-50930905 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem55b | NM_001033271.4 | NP_001028443.1 | ||
AK144251.1 | BAE25798.1 | |||
AK152640.1 | ||||
AK152658.1 | ||||
AK159641.1 | BAE35254.1 | |||
AK169026.1 | 2 | 1432 | ||
AK185482.1 | ||||
AK205319.1 | ||||
AK205696.1 | ||||
AK213231.1 | ||||
AK216843.1 | ||||
AK219909.1 | ||||
BC106163.1 | AAI06164.1 |