Assay Details
Target Gene Details
Entrez Gene ID: | 218756 |
Gene Name: | solute carrier family 4, sodium bicarbonate cotransporter, member 7 |
Gene Aliases: |
E430014N10Rik, NBC3, NBCn1, NBCn1-G |
Location: |
Chr.14:14703011-14799944 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc4a7 | NM_001033270.2 | 6 | 917 | NP_001028442.2 |
AF218295.1 | 2 | 162 | AAF26421.1 | |
AK088400.1 | BAC40330.1 | |||
AK152233.1 | 6 | 918 | BAE31059.1 | |
AK153567.1 | 6 | 918 | BAE32101.1 | |
GU386352.1 | ||||
GU386353.2 | 6 | 863 | ||
HM624050.1 | 6 | 727 | ||
HM624051.1 | ||||
JQ073566.1 | 6 | 709 | ||
JQ344322.1 | ||||
JQ347261.1 | 6 | 727 | ||
JQ349031.1 | 6 | 863 | ||
JQ349032.1 | ||||
JQ349033.1 | ||||
JX254908.1 | 6 | 863 | ||
JX254909.1 | ||||
JX254910.1 | 6 | 863 | ||
JX254911.1 | ||||
JX254912.1 | 6 | 727 | ||
JX310364.1 | ||||
KF279520.1 | 8 | 827 | ||
KF279521.1 |
More Information
Set Membership: |
Intragenic Exonic |