Assay Details
Target Gene Details
Entrez Gene ID: | 76485 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
2410004H05Rik, 5430414N14Rik, AI450005 |
Location: |
Chr.14:31001347-31012441 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt8d1 | NM_001165930.1 | 4 | 333 | NP_001159402.1 |
NM_029626.2 | 4 | 862 | NP_083902.2 | |
AK010388.1 | 4 | 591 | BAB26903.1 | |
AK077388.1 | 4 | 327 | ||
AK160461.1 | 5 | 493 | BAE35802.1 | |
BC055709.1 | 4 | 619 | AAH55709.1 | |
BC058731.1 | 4 | 619 | AAH58731.1 | |
BC069873.1 | 4 | 412 | AAH69873.1 | |
CX209391.1 | 3 | 293 |
More Information
Set Membership: |
Intragenic Exonic |