Assay Details
Target Gene Details
Entrez Gene ID: | 66092 |
Gene Name: | growth hormone inducible transmembrane protein |
Gene Aliases: |
1010001P14Rik, C77840, MICS1, PTD010, Tmbim5 |
Location: |
Chr.14:37120444-37135327 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ghitm | NM_001199122.1 | NP_001186051.1 | ||
NM_078478.5 | NP_510963.1 | |||
AF412297.1 | AAL07803.1 | |||
AK005636.1 | ||||
AK093887.1 | BAC04243.1 | |||
AK134592.1 | BAE22197.1 | |||
AK146433.1 | BAE27167.1 | |||
AK146853.1 | BAE27483.1 | |||
AK151488.1 | BAE30441.1 | |||
AK158904.1 | BAE34723.1 | |||
AK159695.1 | BAE35295.1 | |||
AK159724.1 | BAE35319.1 | |||
AK164122.1 | BAE37638.1 | |||
AK167970.1 | BAE39965.1 | |||
AK168022.1 | BAE40007.1 | |||
AK168678.1 | BAE40527.1 | |||
AK168800.1 | BAE40631.1 | |||
BC008622.1 | AAH08622.1 | |||
BC010224.1 | AAH10224.1 |