Assay Details
Target Gene Details
Entrez Gene ID: | 50905 |
Gene Name: | interleukin 17 receptor B |
Gene Aliases: |
Evi27, IL-17ER, IL-17Rh1, IL17RH1, Il17br |
Location: |
Chr.14:29995506-30008896 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il17rb | NM_019583.3 | 7 | 712 | NP_062529.2 |
AF208108.1 | 7 | 625 | AAF86049.1 | |
AF208109.1 | 7 | 1251 | AAF86050.1 | |
AK045627.1 | 7 | 713 | BAC32438.1 | |
AK050139.1 | 7 | 633 | ||
BC026546.1 | 7 | 612 | AAH26546.1 |
More Information
Set Membership: |
Intragenic Exonic |