Assay Details
Target Gene Details
Entrez Gene ID: | 14702 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 2 |
Gene Aliases: |
82 |
Location: |
Chr.14:19872558-19977462 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gng2 | NM_001038637.1 | NP_001033726.1 | ||
NM_001285908.1 | NP_001272837.1 | |||
NM_001285909.1 | NP_001272838.1 | |||
NM_001285910.1 | NP_001272839.1 | |||
NM_010315.4 | NP_034445.1 | |||
AK003588.1 | BAB22878.1 | |||
AK012405.1 | BAB28219.1 | |||
AK036138.1 | BAC29316.1 | |||
AK075698.1 | BAC35896.1 | |||
AK158697.1 | BAE34615.1 | |||
AK160396.1 | BAE35765.1 | |||
BY108799.1 | ||||
BY245532.1 | ||||
CA324196.1 | ||||
W48309.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |