Assay Details
Target Gene Details
Entrez Gene ID: | 218977 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 5 |
Gene Aliases: |
C77459, C86398, Dap-5, Dlg7, Hurp, mKIAA0008 |
Location: |
Chr.14:47387779-47418407 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlgap5 | NM_144553.2 | 12 | 1761 | NP_653136.2 |
AB076696.1 | 12 | 1761 | BAB97377.1 | |
AK036384.1 | 12 | 1563 | BAC29405.1 | |
AK048470.1 | 12 | 1531 | ||
AK085882.1 | 12 | 1542 | BAC39560.1 | |
AK129032.1 | 12 | 1507 | BAC97842.1 | |
AK135820.1 | 12 | 1568 | BAE22675.1 | |
AK147955.1 | 12 | 1531 | BAE28249.1 | |
BC043924.1 | 4 | 387 | AAH43924.1 | |
BC058087.1 | 12 | 1495 | AAH58087.1 |
More Information
Set Membership: |
Intragenic Exonic |