Assay Details
Target Gene Details
Entrez Gene ID: | 12684 |
Gene Name: | cell death-inducing DNA fragmentation factor, alpha subunit-like effector B |
Gene Aliases: |
1110030C18Rik, AI790179, CIDE-B |
Location: |
Chr.14:55754052-55758424 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cideb | NM_009894.3 | NP_034024.2 | ||
AF041377.1 | AAC34986.1 | |||
AK003968.1 | BAE43153.1 | |||
BC012664.1 | AAH12664.1 | |||
BC046340.1 | AAH46340.1 | |||
CB600615.1 |
Target Gene Details
Entrez Gene ID: | 57260 |
Gene Name: | leukotriene B4 receptor 2 |
Gene Aliases: |
5830462O07Rik, BLT2, LTB4-R 2, LTB4-R-2, LTB4-R2 |
Location: |
Chr.14:55757916-55764066 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltb4r2 |