Assay Details
Target Gene Details
Entrez Gene ID: | 20719 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6a |
Gene Aliases: |
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin |
Location: |
Chr.13:33917918-34002794 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinb6a | NM_001164117.1 | NP_001157589.1 | ||
NM_001164118.1 | NP_001157590.1 | |||
NM_001243192.1 | NP_001230121.1 | |||
NM_009254.3 | NP_033280.1 | |||
AK033731.1 | BAC28452.1 | |||
AK154700.1 | BAE32772.1 | |||
AK167295.1 | BAE39400.1 | |||
AK169145.1 | BAE40924.1 | |||
AK183101.1 | ||||
BC006766.1 | AAH06766.1 | |||
BC057950.1 | AAH57950.1 | |||
CF732441.1 | ||||
CT010346.1 | CAJ18554.1 | |||
U25844.1 | AAA79684.1 |
More Information
Set Membership: |
Intragenic Non-exonic |