Assay Details
Target Gene Details
Entrez Gene ID: | 68750 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
1110037N09Rik, AA414966, B930013M22Rik |
Location: |
Chr.13:37778382-37952005 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rreb1 | NM_001039188.1 | 8 | 1220 | NP_001034277.1 |
NM_001177868.1 | 8 | 945 | NP_001171339.1 | |
NM_001177869.1 | 7 | 881 | NP_001171340.1 | |
NM_026830.2 | 8 | 1188 | NP_081106.1 | |
NR_033218.1 | 8 | 2342 | ||
NR_033615.1 | 8 | 2159 | ||
AK047047.1 | 8 | 2139 | ||
AK047251.1 | 8 | 2141 | ||
AK147260.1 | 8 | 2343 | BAE27803.1 | |
AK147653.1 | 8 | 2343 | BAE28051.1 | |
AK154980.1 | 8 | 1188 | BAE32969.1 | |
AK170652.1 | 8 | 1220 | ||
AK171375.1 | 8 | 925 | BAE42417.1 | |
AY946044.1 | 5 | 572 | AAX83010.1 | |
BC080680.1 | 8 | 1034 | AAH80680.1 |