Assay Details
Target Gene Details
Entrez Gene ID: | 66082 |
Gene Name: | abhydrolase domain containing 6 |
Gene Aliases: |
0610041D24Rik, AA673485, AV065425 |
Location: |
Chr.14:8002807-8056773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd6 | NM_025341.3 | NP_079617.2 | ||
AK002883.1 | BAB22430.1 | |||
AK011887.1 | ||||
AK018260.1 | BAB31136.1 | |||
AK076105.1 | BAC36186.1 | |||
AK090076.1 | BAC41081.1 | |||
AK168782.1 | BAE40616.1 | |||
AK181596.1 | ||||
AK211932.1 | ||||
BC027011.1 | AAH27011.1 |
More Information
Set Membership: |
Intragenic Non-exonic |