Assay Details
Target Gene Details
Entrez Gene ID: | 208366 |
Gene Name: | ribonuclease P 40 subunit |
Gene Aliases: |
D8Bwg1265e, Rnasep1 |
Location: |
Chr.13:35895104-35911565 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpp40 | NM_145938.4 | NP_666050.3 | ||
BC024607.1 | AAH24607.1 |