Assay Details
Target Gene Details
Entrez Gene ID: | 26564 |
Gene Name: | receptor tyrosine kinase-like orphan receptor 2 |
Gene Aliases: |
Ntrkr2, mRor2 |
Location: |
Chr.13:53109317-53286109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ror2 | NM_013846.3 | NP_038874.3 | ||
AB010384.1 | BAA75481.1 | |||
AK031112.1 | BAC27258.1 | |||
AK031789.1 | ||||
AK083538.1 | ||||
AK084752.1 | BAC39273.1 | |||
AK157705.1 | BAE34159.1 | |||
BC030848.1 | AAH30848.1 | |||
BC070436.1 | AAH70436.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |