Assay Details
Target Gene Details
Entrez Gene ID: | 105239 |
Gene Name: | ring finger protein 44 |
Gene Aliases: |
AI854545, mKIAA1100 |
Location: |
Chr.13:54679399-54693995 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf44 | NM_001146025.1 | 5 | 956 | NP_001139497.1 |
NM_001146026.1 | 5 | 881 | NP_001139498.1 | |
NM_001146027.1 | 5 | 747 | NP_001139499.1 | |
NM_134064.2 | 5 | 889 | NP_598825.2 | |
NR_027395.1 | 5 | 809 | ||
NR_027396.1 | 5 | 806 | ||
AK147349.1 | 5 | 882 | BAE27859.1 | |
AK160090.1 | 5 | 740 | BAE35621.1 | |
AK160115.1 | 5 | 740 | BAE35639.1 | |
AK161651.1 | 5 | 869 | ||
AK171943.1 | 5 | 738 | BAE42740.1 | |
AK172257.1 | 5 | 738 | ||
BC017630.1 | 5 | 682 | AAH17630.1 | |
BC035548.1 | 5 | 704 | AAH35548.1 | |
CJ147978.1 |