Assay Details
Target Gene Details
Entrez Gene ID: | 27015 |
Gene Name: | polymerase (DNA directed), kappa |
Gene Aliases: |
Dinb1 |
Location: |
Chr.13:96480689-96542648 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polk | NM_012048.2 | 2 | 209 | NP_036178.1 |
AB027563.1 | 2 | 197 | BAA86942.1 | |
AF163571.1 | 2 | 209 | AAF02541.1 | |
AK042272.1 | 2 | 304 | ||
AK138299.1 | 2 | 212 | BAE23614.1 | |
AY769919.1 | AAV80817.1 | |||
AY769920.1 | AAV80818.1 | |||
AY769921.1 | AAV80819.1 | |||
AY769922.1 | AAV80820.1 | |||
AY769923.1 | AAV80821.1 | |||
AY769924.1 | AAV80822.1 | |||
AY769925.1 | AAV80823.1 | |||
AY769926.1 | AAV80824.1 | |||
AY769927.1 | AAV80825.1 | |||
AY769928.1 | AAV80826.1 | |||
BC052820.1 | 2 | 282 | AAH52820.1 |
More Information
Set Membership: |
Intragenic Exonic |