Assay Details
Target Gene Details
Entrez Gene ID: | 27062 |
Gene Name: | Ca2+-dependent secretion activator |
Gene Aliases: |
AU067781, CAPS, CAPS1, mKIAA1121 |
Location: |
Chr.14:12372560-12823314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cadps | NM_001042617.1 | NP_001036082.1 | ||
NM_012061.3 | NP_036191.2 | |||
AK122453.1 | BAC65735.1 | |||
AK133310.1 | ||||
AK161919.1 | BAE36633.1 | |||
BC057065.1 | AAH57065.1 | |||
D86214.1 | BAA13044.1 |
More Information
Set Membership: |
Intragenic Non-exonic |