Assay Details
Target Gene Details
Entrez Gene ID: | 27015 |
Gene Name: | polymerase (DNA directed), kappa |
Gene Aliases: |
Dinb1 |
Location: |
Chr.13:96480689-96542648 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polk | NM_012048.2 | NP_036178.1 | ||
AB027563.1 | BAA86942.1 | |||
AF163571.1 | AAF02541.1 | |||
AK042272.1 | 1 | 97 | ||
AK138299.1 | BAE23614.1 | |||
BC052820.1 | 1 | 75 | AAH52820.1 |
More Information
Set Membership: |
Intragenic Non-exonic |