Assay Details
Target Gene Details
Entrez Gene ID: | 21810 |
Gene Name: | transforming growth factor, beta induced |
Gene Aliases: |
68kDa, AI181842, AI747162, Beta-ig, big-h3 |
Location: |
Chr.13:56609603-56639339 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfbi | NM_009369.4 | 8 | 1098 | NP_033395.1 |
AK084431.1 | 8 | 1116 | BAC39181.1 | |
AK135536.1 | 8 | 1143 | BAE22571.1 | |
AK142323.1 | 8 | 1118 | BAE25032.1 | |
AK150449.1 | 8 | 1116 | ||
AK150833.1 | 8 | 1116 | ||
AK150969.1 | 8 | 1117 | BAE30000.1 | |
AK151680.1 | 8 | 1113 | BAE30605.1 | |
AK152365.1 | 8 | 1117 | BAE31155.1 | |
AK152662.1 | 8 | 1116 | ||
AK155572.1 | 8 | 1113 | BAE33330.1 | |
AK155828.1 | 8 | 1113 | BAE33452.1 | |
AK159711.1 | 8 | 1112 | ||
AK164373.1 | 8 | 1116 | ||
AK169912.1 | 8 | 1118 | BAE41453.1 | |
AK170495.1 | 8 | 1113 | BAE41834.1 | |
AK171557.1 | 7 | 937 | BAE42525.1 | |
BC129900.1 | 8 | 1091 | ||
BC129901.1 | 8 | 1091 | ||
L19932.1 | 8 | 1099 | AAC37658.1 |
More Information
Set Membership: |
Intragenic Exonic |