Assay Details
Target Gene Details
Entrez Gene ID: | 21810 |
Gene Name: | transforming growth factor, beta induced |
Gene Aliases: |
68kDa, AI181842, AI747162, Beta-ig, big-h3 |
Location: |
Chr.13:56609603-56639339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfbi | NM_009369.4 | NP_033395.1 | ||
AK084431.1 | BAC39181.1 | |||
AK135536.1 | BAE22571.1 | |||
AK142323.1 | BAE25032.1 | |||
AK150449.1 | ||||
AK150833.1 | ||||
AK150969.1 | BAE30000.1 | |||
AK151680.1 | BAE30605.1 | |||
AK152365.1 | BAE31155.1 | |||
AK152662.1 | ||||
AK155572.1 | BAE33330.1 | |||
AK155828.1 | BAE33452.1 | |||
AK159711.1 | ||||
AK164373.1 | ||||
AK170495.1 | BAE41834.1 | |||
AK171557.1 | BAE42525.1 | |||
BC129900.1 | ||||
BC129901.1 | ||||
L19932.1 | AAC37658.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |