Assay Details
Target Gene Details
Entrez Gene ID: | 71037 |
Gene Name: | protease, serine 55 |
Gene Aliases: |
4933401F05Rik |
Location: |
Chr.14:64075435-64085433 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss55 | NM_001081063.1 | 2 | 142 | NP_001074532.1 |
AK016601.1 | 1 | 175 | ||
BC115562.2 | 2 | 143 |
More Information
Set Membership: |
Intragenic Exonic |