Assay Details
Target Gene Details
Entrez Gene ID: | 268686 |
Gene Name: | S100 calcium binding protein, zeta |
Gene Aliases: |
Gm625 |
Location: |
Chr.13:95477266-95479379 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100z | NM_001081159.1 | 2 | 230 | NP_001074628.1 |
BC147382.1 | 2 | 296 | ||
BC147383.1 | 2 | 296 |
More Information
Set Membership: |
Intragenic Exonic |