Assay Details
Target Gene Details
Entrez Gene ID: | 20418 |
Gene Name: | src homology 2 domain-containing transforming protein C3 |
Gene Aliases: |
N-Shc, Rai, ShcC |
Location: |
Chr.13:51431041-51567084 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shc3 | NM_009167.3 | 8 | 681 | NP_033193.2 |
BC103611.1 | 8 | 681 | AAI03612.1 | |
BC103612.1 | 8 | 681 | AAI03613.1 | |
BC105644.1 | 8 | 681 | ||
BC105645.1 | 8 | 681 | ||
U46854.1 | 8 | 681 | AAC52508.1 |
More Information
Set Membership: |
Intragenic Exonic |