Assay Details
Target Gene Details
Entrez Gene ID: | 54373 |
Gene Name: | protease, serine 16 (thymus) |
Gene Aliases: |
AI448615, TSSP |
Location: |
Chr.13:22002176-22009741 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss16 | NM_019429.2 | 9 | 1126 | NP_062302.1 |
AJ131775.1 | 9 | 1126 | CAB66137.1 | |
AK037514.1 | 5 | 1945 | ||
AK040376.1 | 2 | 509 | ||
AK088019.1 | 9 | 1154 | BAC40100.1 | |
BC125481.1 | 9 | 1124 | ||
BC131955.1 | 9 | 1124 |
More Information
Set Membership: |
Intragenic Exonic |