Assay Details
Target Gene Details
Entrez Gene ID: | 18708 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha) |
Gene Aliases: |
PI3K, p50alpha, p55alpha, p85alpha |
Location: |
Chr.13:101680761-101768217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3r1 | NM_001024955.2 | NP_001020126.1 | ||
NM_001077495.2 | NP_001070963.1 | |||
AK046259.1 | ||||
BC026146.1 | AAH26146.1 | |||
BC051106.1 | AAH51106.1 | |||
CB519962.1 | ||||
M60651.1 | AAA39886.1 | |||
U50413.1 | AAC52847.1 | |||
U50414.1 | AAC52848.1 |
More Information
Set Membership: |
Intragenic Non-exonic |