Assay Details
Target Gene Details
Entrez Gene ID: | 105522 |
Gene Name: | ankyrin repeat domain 28 |
Gene Aliases: |
AI465466, AI604979, E430019N21Rik, mKIAA0379 |
Location: |
Chr.14:31698768-31830415 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd28 | NM_001024604.2 | NP_001019775.1 | ||
AK133330.1 | ||||
AK220336.1 | BAD90403.1 | |||
BC051456.1 | AAH51456.1 | |||
BC094609.1 | AAH94609.1 |
More Information
Set Membership: |
Intragenic Non-exonic |