Assay Details
Target Gene Details
Entrez Gene ID: | 108811 |
Gene Name: | coiled-coil domain containing 122 |
Gene Aliases: |
4933415L06Rik |
Location: |
Chr.14:77036772-77112265 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc122 | NM_175369.3 | 1 | 93 | NP_780578.1 |
AK077161.1 | 1 | 94 | BAC36652.1 | |
BC096577.1 | 1 | 87 | AAH96577.1 | |
BC138881.1 | ||||
BC138883.1 |
Target Gene Details
Entrez Gene ID: | 210808 |
Gene Name: | laccase (multicopper oxidoreductase) domain containing 1 |
Gene Aliases: |
9030625A04Rik |
Location: |
Chr.14:77024201-77036898 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lacc1 | AK053693.1 | BAC35476.1 |