Assay Details
Target Gene Details
Entrez Gene ID: | 20706 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 9b |
Gene Aliases: |
1600019A21Rik, R86, SPI-CI, Spi10, ovalbumin |
Location: |
Chr.13:33027414-33040559 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinb9b | NM_011452.2 | 5 | 686 | NP_035582.1 |
AK005491.1 | 5 | 687 | BAB24076.1 | |
AK167563.1 | 5 | 562 | BAE39626.1 | |
AK171850.1 | 5 | 559 | BAE42697.1 | |
BC064768.1 | 5 | 546 | AAH64768.1 | |
U96705.2 | 5 | 687 | AAB57817.2 |
More Information
Set Membership: |
Intragenic Exonic |